missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4,4-difluoropimelate, 97%, Thermo Scientific™

Product Code. 11345155
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345155 1 g 1g
15494137 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345155 Supplier Thermo Scientific Alfa Aesar Supplier No. H31544.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22515-16-8
Molecular Formula C11H18F2O4
Molecular Weight (g/mol) 252.26
MDL Number MFCD08146634
InChI Key XUOBBVMKXUPPEW-UHFFFAOYSA-N
Synonym 4,4-difluoroheptanedioic acid diethyl ester, 1,7-diethyl 4,4-difluoroheptanedioate, diethyl 4,4-difluoropimelate, pubchem22650, diethyl 4,4-heptanedioate, ksc494q0l, diethyl4,4-difluoroheptanedioate, diethyl 4,4-difluoroheptane-1,7-dioate, 4,4-difluoro-heptanedioic acid diethyl ester, heptanedioicacid,4,4-difluoro-,1,7-diethylester
PubChem CID 10264195
IUPAC Name diethyl 4,4-difluoroheptanedioate
SMILES CCOC(=O)CCC(F)(F)CCC(=O)OCC

Specifications

Boiling Point 108°C (3 mmHg)
Refractive Index 1.415
Quantity 1 g
Formula Weight 252.26
Percent Purity 97%
Chemical Name or Material Diethyl 4,4-difluoropimelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.