missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3-Oxopentanedioate, TRC

Product Code. 30372977
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372977 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372977 Supplier Toronto Research Chemicals Supplier No. TRCD4445951G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 3-Oxopentanedioate
CAS 105-50-0
Molecular Formula C9H14O5
Quantity 1 g
Synonym 3-Oxoglutaric Acid Diethyl Ester, beta-Oxoglutaric Acid Diethyl Ester, 1,3-Acetonedicarboxylic Acid Diethyl Ester, 3-Oxopentanedioic Acid Diethyl Ester, Acetonedicarboxylic Acid Diethyl Ester, Diethyl 1,3-Acetonedicarboxylate, Diethyl 2-Oxopropane-1,3-dicarboxylate, Diethyl 3-Ketoglutarate, Diethyl 3-Oxoglutarate, Diethyl 3-Oxopentanedioate, Diethyl Acetonedicarboxylate, Diethyl beta-Ketoglutarate, Diethyl beta-Oxoglutarate, NSC 9013, beta-Ketoglutaric Acid Diethyl Ester
InChI Formula InChI=1S/C9H14O5/c1-3-13-8(11)5-7(10)6-9(12)14-4-2/h3-6H2,1-2H3
SMILES CCOC(=O)CC(=O)CC(=O)OCC
IUPAC Name diethyl 3-oxopentanedioate
Molecular Weight (g/mol) 202.2
Formula Weight 202.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.