missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3-chloropropyl)malonate, 98%

Product Code. 11393676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393676 25 g 25g
11383676 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393676 Supplier Thermo Scientific Alfa Aesar Supplier No. L14776.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18719-43-2 | C10H17ClO4 | 236.692 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18719-43-2
Molecular Formula C10H17ClO4
Molecular Weight (g/mol) 236.692
MDL Number MFCD00009169
InChI Key WUOPYKKJFZHTRA-UHFFFAOYSA-N
Synonym diethyl 3-chloropropyl malonate, diethyl 2-3-chloropropyl malonate, malonic acid, 3-chloropropyl-, diethyl ester, 2-3-chloropropyl malonic acid diethyl ester, acmc-20al9u, diethyl ?-chloropropyl malonate, chloropropylmalonic acid diethyl ester, diethyl 2-3-chloropropyl propanedioate
PubChem CID 140413
IUPAC Name diethyl 2-(3-chloropropyl)propanedioate
SMILES CCOC(=O)C(CCCCl)C(=O)OCC

Specifications

Density 1.101
Boiling Point 102°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.443
Quantity 25 g
Beilstein 1785711
Formula Weight 236.7
Percent Purity 98%
Chemical Name or Material Diethyl (3-chloropropyl)malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.