missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3-chloropropyl)malonate, 98%

Product Code. 11383676
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11383676

Brand: Thermo Scientific Alfa Aesar L14776.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18719-43-2 | C10H17ClO4 | 236.692 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18719-43-2
Molecular Formula C10H17ClO4
Molecular Weight (g/mol) 236.692
MDL Number MFCD00009169
InChI Key WUOPYKKJFZHTRA-UHFFFAOYSA-N
Synonym diethyl 3-chloropropyl malonate, diethyl 2-3-chloropropyl malonate, malonic acid, 3-chloropropyl-, diethyl ester, 2-3-chloropropyl malonic acid diethyl ester, acmc-20al9u, diethyl ?-chloropropyl malonate, chloropropylmalonic acid diethyl ester, diethyl 2-3-chloropropyl propanedioate
PubChem CID 140413
IUPAC Name diethyl 2-(3-chloropropyl)propanedioate
SMILES CCOC(=O)C(CCCCl)C(=O)OCC

Specifications

Density 1.101
Boiling Point 102°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.443
Quantity 5 g
Beilstein 1785711
Formula Weight 236.7
Percent Purity 98%
Chemical Name or Material Diethyl (3-chloropropyl)malonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.