missing translation for 'onlineSavingsMsg'
Learn More

Diethyl-2-propylimidazole-4,5-dicarboxylate, TRC

Product Code. 30384796
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384796 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384796 Supplier Toronto Research Chemicals Supplier No. TRCD44479510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl-2-propylimidazole-4,5-dicarboxylate
CAS 144689-94-1
Molecular Formula C12 H18 N2 O4
Quantity 10 g
Synonym 1H-Imidazole-4,5-dicarboxylic acid, 2-propyl-, 4,5-diethyl ester, 1H-Imidazole-4,5-dicarboxylic acid, 2-propyl-, diethyl ester (9CI), Diethyl 2-propyl-1H-imidazole-4,5-dicarboxylate, Diethyl 2-propylimidazole-4,5-dicarboxylate
InChI Formula InChI=1S/C12H18N2O4/c1-4-7-8-13-9(11(15)17-5-2)10(14-8)12(16)18-6-3/h4-7H2,1-3H3,(H,13,14)
SMILES CCCc1nc(C(=O)OCC)c([nH]1)C(=O)OCC
IUPAC Name diethyl 2-propyl-1H-imidazole-4,5-dicarboxylate
Molecular Weight (g/mol) 254.28
Formula Weight 254.1267
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.