missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-isopentyl-2-methylmalonate, Tech., Thermo Scientific™

Product Code. p-2703862
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 10541123

Brand: Thermo Scientific Maybridge SB00828EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 121823-85-6
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.33
MDL Number MFCD00053035
InChI Key LFMBZEGLMSBWHG-UHFFFAOYSA-N
PubChem CID 233614
IUPAC Name diethyl 2-methyl-2-(3-methylbutyl)propanedioate
SMILES CCOC(=O)C(C)(CCC(C)C)C(=O)OCC

Specifications

Packaging Amber glass bottle
Quantity 10 g
Synonym diethyl 2-isopentyl-2-methylmalonate, diethyl 2-methyl-2-3-methylbutyl propanedioate, 1,3-diethyl 2-methyl-2-3-methylbutyl propanedioate, propanedioic acid,2-methyl-2-3-methylbutyl-, 1,3-diethyl ester, diethyl methyl 3-methylbutyl malonate, zlchem 675, acmc-1c8qr, diethyl2-isopentyl-2-methylmalonate, diethyl 2-methyl-2-3-methylbutyl propane-1,3-dioate, 2-methyl-2-3-methyl-butyl-malonic acid diethyl ester
Formula Weight 244.33
Percent Purity Tech.
Grade Technical
Chemical Name or Material Diethyl 2-isopentyl-2-methylmalonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.