missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-isopentyl-2-methylmalonate, Tech., Thermo Scientific™

Product Code. 10347404
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10347404 100 g 100g
10541123 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347404 Supplier Thermo Scientific Maybridge Supplier No. SB00828FA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 121823-85-6
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.33
MDL Number MFCD00053035
InChI Key LFMBZEGLMSBWHG-UHFFFAOYSA-N
PubChem CID 233614
IUPAC Name diethyl 2-methyl-2-(3-methylbutyl)propanedioate
SMILES CCOC(=O)C(C)(CCC(C)C)C(=O)OCC

Specifications

Quantity 100 g
Synonym diethyl 2-isopentyl-2-methylmalonate, diethyl 2-methyl-2-3-methylbutyl propanedioate, 1,3-diethyl 2-methyl-2-3-methylbutyl propanedioate, propanedioic acid,2-methyl-2-3-methylbutyl-, 1,3-diethyl ester, diethyl methyl 3-methylbutyl malonate, zlchem 675, acmc-1c8qr, diethyl2-isopentyl-2-methylmalonate, diethyl 2-methyl-2-3-methylbutyl propane-1,3-dioate, 2-methyl-2-3-methyl-butyl-malonic acid diethyl ester
Formula Weight 244.33
Percent Purity Tech.
Grade Technical
Chemical Name or Material Diethyl 2-isopentyl-2-methylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.