missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-bromo-2-methylmalonate, 98%

Product Code. 11370114
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11370114

Brand: Thermo Scientific Alfa Aesar L00575.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29263-94-3 | C8H13BrO4 | 253.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29263-94-3
Molecular Formula C8H13BrO4
Molecular Weight (g/mol) 253.09
MDL Number MFCD00009125
InChI Key CSLQAXTUGPUBCW-UHFFFAOYSA-N
Synonym diethyl 2-bromo-2-methylmalonate, diethyl bromomethylmalonate, diethyl methylbromomalonate, 2-bromo-2-methylmalonic acid diethyl ester, 1,3-diethyl 2-bromo-2-methylpropanedioate, 2-bromo-2-methylpropandioic acid diethyl ester, diethyl bromo methyl malonate, acmc-20al99, cslqaxtugpubcw-uhfffaoysa, diethyl-2-bromo-2-methyl-malonate
PubChem CID 94944
IUPAC Name diethyl 2-bromo-2-methylpropanedioate
SMILES CCOC(=O)C(C)(Br)C(=O)OCC

Specifications

Density 1.323
Boiling Point 105°C to 106°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.449
Quantity 25 g
UN Number UN3265
Beilstein 1779673
Formula Weight 253.1
Percent Purity 98%
Chemical Name or Material Diethyl 2-bromo-2-methylmalonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.