missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Acetamidomalonate, TRC

Product Code. 30385687
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385687 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385687 Supplier Toronto Research Chemicals Supplier No. TRCD44313025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Acetamidomalonate
CAS 1068-90-2
Molecular Formula C9 H15 N O5
Quantity 25 g
Synonym Propanedioic acid, 2-(acetylamino)-, 1,3-diethyl ester, Malonic acid, acetamido-, diethyl ester (6CI,7CI,8CI), Propanedioic acid, (acetylamino)-, diethyl ester (9CI), (Acetylamino)malonic acid diethyl ester, (Acetylamino)propanedioic acid diethyl ester, 2-(Acetylamino)propanedioic acid diethyl ester, 2-Acetylamino-malonic acid diethyl ester, Acetamidomalonic acid diethyl ester, Diethyl (N-acetylamino)malonate, Diethyl (acetylamino)malonate, Diethyl 2-(acetamido)propanedioate, Diethyl 2-(acetylamino)propanedioate, Diethyl 2-acetamidomalonate, Diethyl 2-acetylaminomalonate, Diethyl acetamidomalonate, Diethyl α-(acetylamino)malonate, NSC 7645, 2-Acetamidomalonic acid-diethyl ester
InChI Formula InChI=1S/C9H15NO5/c1-4-14-8(12)7(10-6(3)11)9(13)15-5-2/h7H,4-5H2,1-3H3,(H,10,11)
SMILES CCOC(=O)C(NC(=O)C)C(=O)OCC
IUPAC Name diethyl 2-acetamidopropanedioate
Molecular Weight (g/mol) 217.22
Formula Weight 217.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.