missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Acetamido-2-(2-formylethyl) Malonate, TRC

Product Code. 30382190
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382190 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382190 Supplier Toronto Research Chemicals Supplier No. TRCD4431201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 2-Acetamido-2-(2-formylethyl) Malonate
CAS 53908-65-9
Molecular Formula C26 H29 N2 S . I
Quantity 1 g
Synonym 2-(Acetylamino)-2-(3-oxopropyl)-propanedioic Acid 1,3-Diethyl Ester, Acetamido(2-formylethyl)-malonic Acid Diethyl Ester, (Acetylamino)(3-oxopropyl)-propanedioic Acid Diethyl Ester
InChI Formula InChI=1S/C26H29N2S.HI/c1-3-4-5-10-19-28-24-15-8-9-16-25(24)29-26(28)17-11-12-21-18-20-27(2)23-14-7-6-13-22(21)23;/h6-9,11-18,20H,3-5,10,19H2,1-2H3;1H/q+1;/p-1
SMILES [I-].CCCCCCN1\C(=C\C=C\c2cc[n+](C)c3ccccc23)\Sc4ccccc14
IUPAC Name (2Z)-3-hexyl-2-[(E)-3-(1-methylquinolin-1-ium-4-yl)prop-2-enylidene]-1,3-benzothiazole;iodide
Molecular Weight (g/mol) 528.491
Formula Weight 528.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.