missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2,4-dichlorophenyl)malonate, 95%

Product Code. 11491210
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491210 2.5 g 2.5g
11481210 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491210 Supplier Thermo Scientific Alfa Aesar Supplier No. H26330.27
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 111544-93-5 | C13H14Cl2O4 | 305.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111544-93-5
Molecular Formula C13H14Cl2O4
Molecular Weight (g/mol) 305.15
MDL Number MFCD00831103
InChI Key FIGCCTUCURBNIF-UHFFFAOYSA-N
Synonym diethyl 2-2,4-dichlorophenyl malonate, diethyl 2,4-dichlorophenyl malonate, 1,3-diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenylmalonate, 2-2,4-dichloro-phenyl-malonic acid diethyl ester, propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester
PubChem CID 2736112
IUPAC Name diethyl 2-(2,4-dichlorophenyl)propanedioate
SMILES CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl

Specifications

Quantity 2.5 g
Formula Weight 305.16
Percent Purity 95%
Chemical Name or Material Diethyl (2,4-dichlorophenyl)malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.