missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2,4-dichlorophenyl)malonate, 95%

Product Code. 11481210
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 11481210

Brand: Thermo Scientific Alfa Aesar H26330.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111544-93-5 | C13H14Cl2O4 | 305.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111544-93-5
Molecular Formula C13H14Cl2O4
Molecular Weight (g/mol) 305.15
MDL Number MFCD00831103
InChI Key FIGCCTUCURBNIF-UHFFFAOYSA-N
Synonym diethyl 2-2,4-dichlorophenyl malonate, diethyl 2,4-dichlorophenyl malonate, 1,3-diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenylmalonate, 2-2,4-dichloro-phenyl-malonic acid diethyl ester, propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester
PubChem CID 2736112
IUPAC Name diethyl 2-(2,4-dichlorophenyl)propanedioate
SMILES CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl

Specifications

Quantity 500 mg
Formula Weight 305.16
Percent Purity 95%
Chemical Name or Material Diethyl (2,4-dichlorophenyl)malonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.