missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 12-mercaptododecylphosphonate, 95%

Product Code. 15406827
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406827 250 mg 250mg
15496817 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406827 Supplier Thermo Scientific Alfa Aesar Supplier No. H33935.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1049677-30-6 | C16H35O3PS | 338.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1049677-30-6
Molecular Formula C16H35O3PS
Molecular Weight (g/mol) 338.49
MDL Number MFCD11046117
InChI Key JAVFXEXYSKLPHD-UHFFFAOYSA-N
Synonym diethyl 12-mercaptododecylphosphonate, diethyl 12-sulfanyldodecylphosphonate, 12-mercaptododecylphosphonic acid diethyl ester
PubChem CID 45158995
IUPAC Name 12-diethoxyphosphoryldodecane-1-thiol
SMILES CCOP(=O)(CCCCCCCCCCCCS)OCC

Specifications

Linear Formula HS(CH2)12P(O)(OCH2CH3)2
Refractive Index 1.4610-1.4650
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 338.49
Percent Purity 95%
Chemical Name or Material Diethyl 12-mercaptododecylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.