missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-octylphosphonate, 98%

Product Code. 11379425
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379425 5 g 5g
11389425 25 g 25g
11399425 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379425 Supplier Thermo Scientific Alfa Aesar Supplier No. L18588.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1068-07-1 | C12H27O3P | 250.319 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-07-1
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.319
MDL Number MFCD00015659
InChI Key MVDVRXOWIPONFY-UHFFFAOYSA-N
Synonym diethyl octylphosphonate, diethyl 1-octylphosphonate, phosphonic acid, octyl-, diethyl ester, acmc-1bsbo, octylphosphonic acid diethyl ester, diethyl 1-octylphosphonate, 98+%, octyl-phosphonic acid diethyl ester, phosphonic acid,p-octyl-, diethyl ester, phosphonic acid, p-octyl-, diethyl ester
PubChem CID 70604
IUPAC Name 1-diethoxyphosphoryloctane
SMILES CCCCCCCCP(=O)(OCC)OCC

Specifications

Density 0.953
Boiling Point 88°C to 89°C (0.1 mmHg)
Refractive Index 1.434
Quantity 5 g
Beilstein 1778018
Formula Weight 250.32
Percent Purity ≥98%
Chemical Name or Material Diethyl 1-octylphosphonate
compliance-icons
Product Identifier
  • Diethyl 1-octylphosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Diethyl octylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.