missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-naphthylmethylphosphonate, 90+%

Product Code. 11367375
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367375 50 g 50g
11357375 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367375 Supplier Thermo Scientific Alfa Aesar Supplier No. L05286.18
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 53575-08-9 | C15H19O3P | 278.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53575-08-9
Molecular Formula C15H19O3P
Molecular Weight (g/mol) 278.288
MDL Number MFCD00015769
InChI Key AFNZPMQIKMSUTD-UHFFFAOYSA-N
Synonym diethyl 1-naphthylmethylphosphonate, 1-naphthylmethyl phosphonic acid diethyl ester, diethyl naphthalen-1-ylmethyl phosphonate, diethyl 1-naphthylmethyl phosphonate, phosphonic acid, 1-naphthalenylmethyl-, diethyl ester, diethyl naphthalen-1-ylmethylphosphonate, phosphonic acid, 1-naphthylmethyl-, diethyl ester, 1-diethoxyphosphorylmethyl naphthalene, acmc-1aoev
PubChem CID 101271
IUPAC Name 1-(diethoxyphosphorylmethyl)naphthalene
SMILES CCOP(=O)(CC1=CC=CC2=CC=CC=C21)OCC

Specifications

Boiling Point 205°C to 206°C (5 mmHg)
Refractive Index 1.561
Quantity 50 g
Beilstein 2943656
Formula Weight 278.29
Percent Purity 95%
Chemical Name or Material Diethyl 1-naphthylmethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.