missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate, 97%, Thermo Scientific™

Product Code. 10242982
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10242982 10 g Amber Glass Bottle 10g
10560922 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10242982 Supplier Thermo Scientific Maybridge Supplier No. KM09423EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17740-40-8
Molecular Formula C17H23NO4
Molecular Weight (g/mol) 305.374
MDL Number MFCD00830964
InChI Key LDUSEIANLSWKPY-UHFFFAOYSA-N
Synonym 2,5-diethyl 1-benzylpyrrolidine-2,5-dicarboxylate, 1-benzyl-pyrrolidine-2,5-dicarboxylic acid diethyl ester, diethyl meso-1-benzyl-2,5-pyrrolidinedicarboxylate, 1-benzyl-2,5-bis ethoxycarbonyl pyrrolidine, diethyl-1-benzylpyrrolidine-2,5-dicarboxylate, pubchem15289, diethyl1-benzylpyrrolidine-2,5-dicarboxylate, hcl, diethyl 1-phenylmethyl pyrrolidine-2,5-dicarboxylate, 1-phenylmethyl pyrrolidine-2,5-dicarboxylic acid diethyl ester
PubChem CID 286120
IUPAC Name diethyl 1-benzylpyrrolidine-2,5-dicarboxylate
SMILES CCOC(=O)C1CCC(N1CC2=CC=CC=C2)C(=O)OCC

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 305.37
Percent Purity 95%
Chemical Name or Material Diethyl 1-benzylpyrrolidine-2, 5-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.