missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylvinylsilane, 97%

Product Code. 11369055
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369055 100 g 100g
11349055 5 g 5g
11359055 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369055 Supplier Thermo Scientific Alfa Aesar Supplier No. L14224.22
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5507-44-8 | C7H16O2Si | 160.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5507-44-8
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009067
InChI Key MBGQQKKTDDNCSG-UHFFFAOYSA-N
Synonym diethoxy methyl vinyl silane, methylvinyldiethoxysilane, silane, ethenyldiethoxymethyl, vinylmethyldiethoxysilane, diethoxy methyl vinylsilane, silane, diethoxymethylvinyl, vinyldiethoxymethylsilane, vinylmethyldiathoxysilan, diethoxymethylvinyl silane, diethoxy-methyl-vinyl-silane
PubChem CID 79646
IUPAC Name ethenyl-diethoxy-methylsilane
SMILES CCO[Si](C)(C=C)OCC

Specifications

Boiling Point 133°C to 134°C
Quantity 100 g
Flash Point 17°C (62°F)
Refractive Index 1.4
Sensitivity Moisture sensitive
Density 0.858
UN Number UN1993
Beilstein 970124
Formula Weight 160.29
Percent Purity 97%
Chemical Name or Material Diethoxymethylvinylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.