missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylsilane, 96%

Product Code. 11479422
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479422 10 g 10g
11334509 5 g 5g
11489422 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479422 Supplier Thermo Scientific Alfa Aesar Supplier No. A10153.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2031-62-1 | C5H13O2Si | 133.242 g/mol

Diethoxymethylsilane is used in the preparation of N,N'-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethoxymethylsilane is used in the preparation of N,N′-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.

Solubility
Miscible with water, 1,4-dioxane, tetrahydrofuran, acetonitrile, dichloromethane, chloroform, dimethyl sulfoxide, hexamethylphosphoric triamide and toluene. Insoluble in diethyl ether and hexane.

Notes
Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2031-62-1
Molecular Formula C5H13O2Si
Molecular Weight (g/mol) 133.242
MDL Number MFCD00026749
InChI Key GAURFLBIDLSLQU-UHFFFAOYSA-N
Synonym diethoxymethylsilane, methyldiethoxysilane, diethoxy methyl silane, silane, diethoxymethyl, diethoxy-methylsilane, diethoxy methyl silicon, diethoxymethylsilane. methylhydrogendiethoxysilane., diethoxymethyl-silan, diethoxy methyl silyl
PubChem CID 6327387
IUPAC Name diethoxy(methyl)silicon
SMILES CCO[Si](C)OCC

Specifications

Boiling Point 94°C to 95°C
Quantity 10 g
Linear Formula CH3SiH(OCH2CH3)2
UN Number UN1993
Beilstein 1733975
Flash Point 10°C (50°F)
Solubility Information Miscible with water,1,4-dioxane,tetrahydrofuran,acetonitrile,dichloromethane,chloroform,dimethyl sulfoxide,hexamethylphosphoric triamide and toluene. Insoluble in diethyl ether and hexane.
Formula Weight 134.25
Percent Purity 96%
Refractive Index 1.376
Sensitivity Moisture sensitive
Density 0.84
Chemical Name or Material Diethoxymethylsilane
Show More Show Less

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.