missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethane, 99%, pure, stabilized

Product Code. 10432451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
Unit Size:
500mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432451 500 mL Glass bottle 500mL
10023460 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432451 Supplier Thermo Scientific Acros Supplier No. 296455000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 462-95-3 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-95-3 , 128-37-0
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00009246
InChI Key KLKFAASOGCDTDT-UHFFFAOYSA-N
Synonym diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
PubChem CID 10024
IUPAC Name ethoxymethoxyethane
SMILES CCOCOCC

Specifications

Melting Point -66.5°C
Color Colorless
Density 0.8310g/mL
Boiling Point 87.5°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2OCH2OCH2CH3
Refractive Index 1.3720 to 1.3740
Quantity 500 mL
Specific Gravity 0.831
Solubility Information Solubility in water: 4.2%. Other solubilities: miscible with ethanol, ethyl ether, very soluble in acetone, soluble in benzene, chloroform
Stabilizer 100ppm BHT max.
Viscosity 0.405 mPa.s (20°C)
Formula Weight 104.15
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Diethoxymethane, 99%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.