missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethane, 99.7%, Honeywell™

Product Code. 15665810 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2 L
2.5 L
Packaging:
Glass bottle
Unit Size:
2 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15665810 2.5 L Glass bottle 500mL
15655810 2 L Glass bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15665810 Supplier Honeywell Research Chemicals Supplier No. 538280500ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contains 50-150 ppm BHT as stabilizer, 99.7%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-95-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00009246
InChI Key KLKFAASOGCDTDT-UHFFFAOYSA-N
Synonym diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
PubChem CID 10024
IUPAC Name (ethoxymethoxy)ethane
SMILES CCOCOCC

Specifications

Melting Point -50°C
Color Colorless
Density 0.831g/mL at 25°C
Boiling Point 255°C
Packaging Glass bottle
Linear Formula CH2(OC2H5)2
Quantity 2.5 L
UN Number UN2373
Beilstein 1697253
Vapor Density 3.6 (vs air)
Formula Weight 104.15g/mol
Vapor Pressure 60mmHg (25°C)
Percent Purity 0.97
Chemical Name or Material Diethoxymethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.