missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydimethylsilane, 97%

Product Code. 11373656
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373656 1000 g 1kg
11353656 50 g 50g
11363656 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373656 Supplier Thermo Scientific Alfa Aesar Supplier No. L14725.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-62-6 | C6H16O2Si | 148.277 g/mol

Diethoxydimethylsilane is useful for blocking hydroxyl and amino groups in organic synthesis reactions. This silylating step allows subsequent reactions to be carried out which would be adversely affected by the presence of active hydrogen in the hydroxyl or amine groups. Following the reaction step, hydroxyl or amine groups blocked with diethoxydimethylsilane may be recovered by a hydrolysis procedure. It is also used for preparing hydrophobic and release materials as well as enhancing flow of powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethoxydimethylsilane is useful for blocking hydroxyl and amino groups in organic synthesis reactions. This silylating step allows subsequent reactions to be carried out which would be adversely affected by the presence of active hydrogen in the hydroxyl or amine groups. Following the reaction step, hydroxyl or amine groups blocked with diethoxydimethylsilane may be recovered by a hydrolysis procedure. It is also used for preparing hydrophobic and release materials as well as enhancing flow of powders.

Solubility
Reacts with water.

Notes
Store under dry inert gas. It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-62-6
Molecular Formula C6H16O2Si
Molecular Weight (g/mol) 148.277
MDL Number MFCD00009068
InChI Key YYLGKUPAFFKGRQ-UHFFFAOYSA-N
Synonym dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22
PubChem CID 62322
IUPAC Name diethoxy(dimethyl)silane
SMILES CCO[Si](C)(C)OCC

Specifications

Boiling Point 111°C to 113°C
Melting Point -70°C
Quantity 1000 g
UN Number UN2380
Beilstein 1736110
Flash Point 11°C (52°F)
Solubility Information Reacts with water.
Formula Weight 148.28
Percent Purity 97%
Odor Mild
Refractive Index 1.382
Sensitivity Moisture sensitive
Density 0.865
Chemical Name or Material Diethoxydimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.