missing translation for 'onlineSavingsMsg'
Learn More

Diethazine Sulfoxide N-Oxide Hydrochloride (Diethazine N,S-Dioxide Hydrochloride), Mikromol™

Product Code. 30052213
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30052213 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30052213

Brand: Mikromol MM1593.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diethazine Sulfoxide N-Oxide Hydrochloride (Diethazine N,S-Dioxide Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethazine Sulfoxide N-Oxide Hydrochloride
Analyte or Component Names Diethazine Sulfoxide N-Oxide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 O2 S . Cl H
Certifications/Compliance ISO 9001
Synonym Diethazine Sulfoxide N-Oxide Hydrochloride
InChI Formula InChI=1S/C18H22N2O2S.ClH/c1-3-20(21,4-2)14-13-19-15-9-5-7-11-17(15)23(22)18-12-8-6-10-16(18)19;/h5-12H,3-4,13-14H2,1-2H3;1H
SMILES Cl.CC[N+]([O-])(CC)CCN1c2ccccc2S(=O)c3ccccc13
IUPAC Name N,N-diethyl-2-(5-oxophenothiazin-10-yl)ethanamine oxide;hydrochloride
Molecular Weight (g/mol) 366.91
Quantity 25 mg
Formula Weight 366.1169 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.