missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, BAKER ANALYZED™, J.T. Baker™

Product Code. 15588384 Shop All J.T.Baker Products
Change view
Click to view available options
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15588384 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588384 Supplier JT Baker Supplier No. 7026.2500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratory Reagent

Specifications

CAS 111-42-2
Melting Point 28°C
Density 1.1 g/cm³ (20°C)
Boiling Point 268.8°C (1013 hPa)
Flash Point 176°C
Ignition Residue max. 0.005%
Assay Percent Range min. 98.5%
Packaging Glass bottle
Molecular Formula C4H11NO2
MDL Number MFCD00002843
Quantity 2.5 L
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
SMILES OCCNCCO
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
Molecular Weight (g/mol) 105.14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.