missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, 99%

Product Code. p-3736221 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
Glass Bottle
Metal drum
Unit Size:
1kg
2.5kg
25kg
This item is not returnable. View return policy

Product Code. 10487100

Brand: Thermo Scientific Acros 113920010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO

Specifications

Density 1.0955g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (HOCH2CH2)2NH
Refractive Index 1.4760 to 1.4790 (20°C, 589nm)
Quantity 1 kg
Beilstein 04, 283
Merck Index 15, 3118
Formula Weight 105.14
Vapor Pressure 2hPa at 10°C
Percent Purity ≥98.5%
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Diethanolamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethanolamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.