missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diesel Fuel 2 Composite 50000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Spezifikation
Spezifikation
| Chemical Name or Material | No. 2 Diesel Oil |
| CAS | 68334-30-5 |
| Analyte or Component Names | No. 2 Diesel Oil |
| Shipping Condition | Room Temperature |
| Concentration | 50000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Fuels, diesel, Diesel fuel oil, Diesel oil, Diesel oil (petroleum), Diesel test fuel, HRD 76, NATO-F 76, Diesel fuel |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?