missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dienogest Antibody (C11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43740
This item is not returnable.
View return policy
Description
Dienogest Monoclonal antibody specifically detects Dienogest in Nonspecies specific samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| Dienogest | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunohistochemistry-Frozen 5-20 μg/mL | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Non-species specific | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| C11 | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| BSA - Dienogest | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction