missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DIDS
GREENER_CHOICE

Product Code. 18004002
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18004002

Brand: Tocris Bioscience™ 4523/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chloride channel blocker; TRPV1 modulator; RAD51 recombinase inhibitor

ClC-Ka chloride channel blocker (IC50 = 100 μM). Blocks the maxi chloride channel in apical membranes from human placenta. Displays antagonistic activity at TRPM4 and TRPC4 channels; potentiates agonist-induced TRPV1 currents (IC50 = 4.88 μM in rat DRG neurons). Inhibits RAD51 recombinase activity (KD = 2 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 67483-13-0
Quantity 50 mg
Molecular Formula C16H8N2Na2O6S4
Formula Weight 498.48
Purity >95%
Molecular Weight (g/mol) 498.468
Chemical Name or Material DIDS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.