missing translation for 'onlineSavingsMsg'
Learn More

Didox, MedChemExpress

Product Code. 16477758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477758 10 mg 10mg
16473727 50 mg 50mg
16437616 100 mg 100mg
16410228 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477758 Supplier Medchem Express Supplier No. HY19387
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Didox (NSC-324360) is a synthetic ribonucleotide reductase (RR) inhibitor.

Didox (NSC-324360) is a synthetic ribonucleotide reductase (RR) inhibitor.

Specifications

Chemical Name or Material Didox
CAS 69839-83-4
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C7H7NO4
Quantity 10 mg
Synonym NSC-324360
Solubility Information DMSO : 100 mg/mL (591.26 mM; Need ultrasonic)
SMILES O=C(NO)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 169.13
Formula Weight 169.13
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.