missing translation for 'onlineSavingsMsg'
Learn More

Didodecylmethylamine, TRC

Product Code. 30382690
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382690 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382690 Supplier Toronto Research Chemicals Supplier No. TRCD4417351G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Didodecylmethylamine
CAS 2915-90-4
Molecular Formula C25 H53 N
Quantity 1 g
Synonym N-Dodecyl-N-methyl-1-dodecanamine, Dilaurylmethylamine, Dilaurylmonomethylamine, Farmin M 2-2095, Farmin M2 20, Methyldidodecylamine, N,N-Didodecyl-N-methylamine, N,N-Didodecylmethylamine, N-Dodecyl-N-methyldodecan-1-amine, N-Methyl-N,N-didodecylamine, N-Methyldidodecylamine, N-Dodecyl-N-methyldodecan-1-amine
InChI Formula InChI=1S/C25H53N/c1-4-6-8-10-12-14-16-18-20-22-24-26(3)25-23-21-19-17-15-13-11-9-7-5-2/h4-25H2,1-3H3
SMILES CCCCCCCCCCCCN(C)CCCCCCCCCCCC
IUPAC Name N-dodecyl-N-methyldodecan-1-amine
Molecular Weight (g/mol) 367.695
Formula Weight 367.418
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.