missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Didodecyldimethylammonium Chloride-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Didodecyldimethylammonium Chloride-d6 |
| Molecular Formula | C26H50D6ClN |
| Quantity | 50 mg |
| Synonym | N-Dodecyl-N,N-dimethyl-1-dodecanaminium Chloride-d6, Q 212-d6, Aliquat 204-d6, Dilauryldimethylammonium Chloride-d6, Dimethyldidodecylammonium Chloride-d6, Dimethyldilaurylammonium Chloride-d6, Isothan DL-d6, Isothan DL 1-d6, Quartamin D 24E-d6, Quaternium 47-d6 |
| InChI Formula | InChI=1S/C26H56N.ClH/c1-5-7-9-11-13-15-17-19-21-23-25-27(3,4)26-24-22-20-18-16-14-12-10-8-6-2;/h5-26H2,1-4H3;1H/q+1;/p-1/i3D3,4D3; |
| SMILES | CCCCCCCCCCCC[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])CCCCCCCCCCCC.[Cl-] |
| Molecular Weight (g/mol) | 424.22 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?