missing translation for 'onlineSavingsMsg'
Learn More

Didodecyldimethylammonium bromide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313845
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313845 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313845 Supplier Dr. Ehrenstorfer Supplier No. DREB12588350AL100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Didodecyldimethylammonium Bromide
CAS 3282-73-3
Analyte or Component Names Didodecyldimethylammonium Bromide
Shipping Condition +20°C
Molecular Formula C26H56BrN
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1-Dodecanaminium, N-dodecyl-N,N-dimethyl-, bromide (9CI), Ammonium, didodecyldimethyl-, bromide (8CI), Didodecyldimethylammonium bromide (7CI), Bis(dodecyl)dimethylammonium bromide, Bis(n-dodecyl)dimethylammonium bromide, DDAB, DDDMA, DMAB, Di-n-dodecyldimethylammonium bromide, Dilauryldimethylammonium bromide, Dimethyldidodecylammonium bromide, Dimethyldilaurylammonium bromide
InChI Formula InChI=1S/C26H56N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27(3,4)26-24-22-20-18-16-14-12-10-8-6-2;/h5-26H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC
IUPAC Name didodecyl(dimethyl)ammonium;bromide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 462.63
Quantity 10 mL
Formula Weight 461.3596 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.