missing translation for 'onlineSavingsMsg'
Learn More

Didodecyl phthalate, 97%

Product Code. 10635671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10635671 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10635671 Supplier Thermo Scientific Acros Supplier No. 407130250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2432-90-8 | C32H54O4 | 502.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2432-90-8
Melting Point 21.0°C to 23.0°C
Color Yellow to Tan
Density 0.9400g/mL
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C32H54O4
Refractive Index 1.4800 to 1.4820
MDL Number MFCD00041920
Quantity 25 g
Beilstein 09, IV, 3186
Specific Gravity 0.94
Synonym didodecyl phthalate, dilauryl phthalate, didodecylphthalate, 1,2-benzenedicarboxylic acid, didodecyl ester, di-n-dodecyl phthalate, phthalic acid, didodecyl ester, 1,2-benzenedicarboxylicacid, 1,2-didodecyl ester, 1,2-benzenedicarboxylic acid, 1,2-didodecyl ester, 1, didodecyl ester, phthalic acid bis-dodecyl ester
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, diethyl ether, benzene
InChI Key PUFGCEQWYLJYNJ-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCCCC
IUPAC Name didodecyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 502.77
PubChem CID 17082
Formula Weight 502.77
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material Didodecyl phthalate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.