missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIDO1, Mouse anti-Human, Clone: 3B1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00011083-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: ILQVQDETHSETADQQEAKWRPGDADGTDCTSIGTIEQKSSEDQGIKGRIEKAANPSGKKKLKIFQPVIEAPGASKCIGPGCCHVAQPDSVYCSNDCILKSpecifications
| DIDO1 | |
| Monoclonal | |
| Unconjugated | |
| death inducer-obliterator 1 | |
| BYE1/C20orf158/DATF1/DIDO2/DIDO3/DIO-1/DIO1/DKFZp434P1115/FLJ11265/KIAA0333/MGC16140/dJ885L7.8 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3B1 | |
| In 1x PBS, pH 7.4 | |
| BC014489 | |
| DIDO1 | |
| DIDO1 (AAH14489, 321 a.a. ∼ 420 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 11083 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction