missing translation for 'onlineSavingsMsg'
Learn More

DIDO1 Antibody [DyLight 650], Novus Biologicals™

Product Code. 30738317 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738317 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738317 Supplier Novus Biologicals Supplier No. AF6947W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DIDO1 Polyclonal antibody specifically detects DIDO1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIDO1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias BYE1, C20orf158, DATF-1, DATF1DIDO3, death associated transcription factor 1, death inducer-obliterator 1, Death-associated transcription factor 1, death-inducer obliterator 1, DIO1DIDO2, DIO-1hDido1, dJ885L7.8, DKFZp434P1115, FLJ11265, KIAA0333chromosome 20 open reading frame 158, MGC16140
Host Species Sheep
Immunogen E. coli-derived recombinant human DIDO1, Met1-Ser117, Accession # Q9BTC0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 11083
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.