missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIDO1 Antibody (3B1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00011083-M04
This item is not returnable.
View return policy
Description
DIDO1 Monoclonal antibody specifically detects DIDO1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
Specifications
| DIDO1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| BYE1, C20orf158, DATF-1, DATF1DIDO3, death associated transcription factor 1, death inducer-obliterator 1, Death-associated transcription factor 1, death-inducer obliterator 1, DIO1DIDO2, DIO-1hDido1, dJ885L7.8, DKFZp434P1115, FLJ11265, KIAA0333chromosome 20 open reading frame 158, MGC16140 | |
| DIDO1 (AAH14489, 321 a.a. ~ 420 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.ILQVQDETHSETADQQEAKWRPGDADGTDCTSIGTIEQKSSEDQGIKGRIEKAANPSGKKKLKIFQPVIEAPGASKCIGPGCCHVAQPDSVYCSNDCILK | |
| 0.1 mg | |
| Apoptosis | |
| 11083 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA | |
| 3B1 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
| AAH14489 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction