missing translation for 'onlineSavingsMsg'
Learn More

Didesmethylchlorpromazine Hydrochloride, TRC

Product Code. 30376467
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376467 100 mg
30378049 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376467 Supplier Toronto Research Chemicals Supplier No. TRCD440905100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3763-80-2
Molecular Formula C15H16Cl2N2S
Molecular Weight (g/mol) 327.27
Synonym 10-(3-Aminopropyl)-2-chloro-phenothiazine Monohydrochloride, Didemethylchlorpromazine Hydrochloride, Didesmethylchlorpromazine Hydrochloride, SKF 4577A
SMILES Cl.NCCCN1c2ccccc2Sc3ccc(Cl)cc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H15ClN2S.ClH/c16-11-6-7-15-13(10-11)18(9-3-8-17)12-4-1-2-5-14(12)19-15;/h1-2,4-7,10H,3,8-9,17H2;1H
Formula Weight 326.0411
Chemical Name or Material Didesmethylchlorpromazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.