missing translation for 'onlineSavingsMsg'
Learn More

Didesmethyl Erlotinib Hydrochloride Salt, TRC

Product Code. 30378912
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378912 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378912 Supplier Toronto Research Chemicals Supplier No. TRCD4411051MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 183320-12-9
Molecular Formula C20 H19 N3 O4 . Cl H
Quantity 1 mg
Synonym Ethanol, 2-[[4-[(3-ethynylphenyl)amino]-6-(2-hydroxyethoxy)-7-quinazolinyl]oxy]-, hydrochloride (1:1), Ethanol, 2,2'-[[4-[(3-ethynylphenyl)amino]-6,7-quinazolinediyl]bis(oxy)]bis-, monohydrochloride (9CI), 2,2'-[[4-[(3-Ethynylphenyl)amino]-6,7-quinazolinediyl]bis(oxy)]bis[ethanol] hydrochloride, CP 373943 HCl, Didesmethyl Erlotinib hydrochloride, Didesmethylerlotinib hydrochloride, N-(3-Ethynylphenyl)-6,7-bis(2-hydroxyethoxy)-4-quinazolinamine hydrochloride
InChI Formula InChI=1S/C20H19N3O4.ClH/c1-2-14-4-3-5-15(10-14)23-20-16-11-18(26-8-6-24)19(27-9-7-25)12-17(16)21-13-22-20;/h1,3-5,10-13,24-25H,6-9H2,(H,21,22,23);1H
SMILES Cl.OCCOc1cc2ncnc(Nc3cccc(c3)C#C)c2cc1OCCO
IUPAC Name 2-[4-(3-ethynylanilino)-7-(2-hydroxyethoxy)quinazolin-6-yl]oxyethanol;hydrochloride
Molecular Weight (g/mol) 401.84
Formula Weight 401.1142
Chemical Name or Material Didesmethyl Erlotinib Hydrochloride Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.