missing translation for 'onlineSavingsMsg'
Learn More

Didesethyl Chloroquine Hydroxyacetamide-d4, TRC

Product Code. 30387317
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387317 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30387317 Supplier Toronto Research Chemicals Supplier No. TRCD44098210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Didesethyl Chloroquine Hydroxyacetamide 1,1,2,2-pentyl-d4
CAS 1216956-86-3
Molecular Formula C16 D4 H16 Cl N3 O2
Quantity 10 mg
Synonym 4-(4'-Glycolamidyl-1'-methyl-3,4-d4-butylamino)-7-chloroquinoline
InChI Formula InChI=1S/C16H20ClN3O2/c1-11(3-2-7-19-16(22)10-21)20-14-6-8-18-15-9-12(17)4-5-13(14)15/h4-6,8-9,11,21H,2-3,7,10H2,1H3,(H,18,20)(H,19,22)/i2D2,7D2
SMILES [2H]C([2H])(CC(C)Nc1ccnc2cc(Cl)ccc12)C([2H])([2H])NC(=O)CO
IUPAC Name N-[4-[(7-chloro-4-quinolyl)amino]-1,1,2,2-tetradeuterio-pentyl]-2-hydroxy-acetamide
Molecular Weight (g/mol) 325.83
Formula Weight 325.1495
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.