missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Didecyl Dimethyl Ammonium-d6 Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Didecyl Dimethyl Ammonium-d6 Chloride |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22H42D6ClN |
| Quantity | 5 mg |
| Synonym | N-Decyl-N,N-dimethyl-1-decanaminium Chloride-d6, Didecyldimethyl-ammonium Chloride-d6, Didecyldimethylammonium Chloride-d6, AQ 210-d6, Acticide DDQ-d6, Acticide DDQ 40-d6, Arquad 10-d6, Arquad 210-d6, Arquad 210-50-d6, Arquad 210-50E-d6, Arquad 210-80-d6, Arquad 210-80E-d6, Arquad 210-85E-d6, Asepas 3-d6, Astop-d6, BTC 1010-d6, BTC 2250-d6, BTC 99-d6, Bardac 22-d6, Bardac 2240v, Bardac 2250-d6, Bardac 2270E-d6, Bardac 2280-d6, Bio-Dac-d6, Britewood Q-d6, Britewood XL-d6, Calgon H 130-d6, Catiogen DDM-d6, Catiogen DDM-PG-d6, Cation DDC-d6, Cation DDC 50-d6, D 10P-d6, DDAC-d6, DDC 80-d6, Dodigen 1881-d6, H 130-d6, H 130 (molluscicide)-d6, K-Sanit BP 80-d6, Kamin RM 2D50A-d6, KleenGrow, M 21080-d6, Macrotrol MT 200-d6, Maquat 4480E-d6, Microbiocide B 74-d6, Nissan Cation 2DB-d6, Nissan Cation 2DB500E-d6, Quartamin D 10E-d6, Quartamin D 10P-d6, Quaternium 12-d6, Querton 210CL-d6, Slaoff 91-d6, Sporekill-d6, Stenquat 1010-d6, Timbercote 2000-d6, Tret-O-Lite XC 507-d6 |
| InChI Formula | InChI=1S/C22H48N.ClH/c1-5-7-9-11-13-15-17-19-21-23(3,4)22-20-18-16-14-12-10-8-6-2;/h5-22H2,1-4H3;1H/q+1;/p-1/i3D3,4D3; |
| SMILES | CCCCCCCCCC[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])CCCCCCCCCC.[Cl-] |
| Molecular Weight (g/mol) | 368.11 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?