missing translation for 'onlineSavingsMsg'
Learn More

Didecyl Dimethyl Ammonium Chloride, TRC

Product Code. 30012343
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012343 25 g 25g
30010267 10 g 10g
30008989 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012343 Supplier Toronto Research Chemicals Supplier No. TRCD43937525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7173-51-5
Molecular Formula C22 H48 N . Cl
Molecular Weight (g/mol) 362.08
Synonym 1-Decanaminium, N-decyl-N,N-dimethyl-, chloride (9CI), Ammonium, didecyldimethyl-, chloride (8CI), Didecyldimethylammonium chloride (7CI), 2DB500E, AQ 210, Acticide, Acticide DDQ, Acticide DDQ 40, Arquad 10, Arquad 210, Arquad 210-50, Arquad 210-50E, Arquad 210-80, Arquad 210-80E, Arquad 210-85E, Asepas 3, Astop, BTC 1010, BTC 1010E, BTC 2250, BTC 99, Bardac 22, Bardac 2240, Bardac 2250, Bardac 2250J, Bardac 2250R, Bardac 2270E, Bardac 2270ETM, Bardac 2280, Bardac 2280G, Bardac 2280R, Bardac 50, Bio-Dac, Britewood Q, Britewood XL, Calgon H 130, Catiogen DDM, Catiogen DDM-PG, Cation DDC, Cation DDC 50, Cation DDC 80, D 10P, DDAC, DDC 80, Didecyldimethylammounium chloride, Dimethyldidecylammonium chloride, Dodigen 1881, Fentacare 1021-80, H 130, H 130 (molluscicide), K-Sanit BP 80, Kamin RM 2D50A, KleenGrow, Lipoquad 210-80E, M 21080, Macrotrol MT 200, Maquat 4450T, Maquat 4480E, Microbiocide B 74, Microbiocide N 750, N,N-Didecyl-N,N-dimethylammonium chloride, New Des 50, Nissan Cation 2DB, Nissan Cation 2DB500E, Nissan Cation 2DB800E, Quartamin D 10E, Quartamin D 10P, Quaternium 12, Querton 210CL, Septapav KhS 70, Slaoff 91, Sporekill, Stenquat 1010, Timbercote 2000, Tret-O-Lite XC 507
IUPAC Name didecyl(dimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCC[N+](C)(C)CCCCCCCCCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C22H48N.ClH/c1-5-7-9-11-13-15-17-19-21-23(3,4)22-20-18-16-14-12-10-8-6-2;/h5-22H2,1-4H3;1H/q+1;/p-1
Formula Weight 361.3475
Percent Purity >95%
Chemical Name or Material Didecyldimethylammonium chloride (DDAC-C10)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.