missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentenyl acrylate

Product Code. 15422787
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15422787

Brand: Thermo Scientific Alfa Aesar 046864.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33791-58-1 | C13H17O2 | 205.28 g/mol

Dicyclopentenyl acrylate is used in organic synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33791-58-1
Molecular Formula C13H17O2
Molecular Weight (g/mol) 205.28
MDL Number MFCD00171305
Synonym DCPA; Tricyclo[5.2.1.02.6]dec-4-enyl acrylate

Specifications

Melting Point -40°C
Color Brown
Density 1.07 g/mL
Flash Point 131°C (268°F)
Quantity 25 g
Solubility Information Fully miscible in water.
Formula Weight 204.27
Physical Form Liquid
Chemical Name or Material Dicyclopentenyl acrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.