missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentamethylenethiuram Disulfide, TRC

Product Code. 30387108
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387108 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387108 Supplier Toronto Research Chemicals Supplier No. TRCD4392201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dicyclopentamethylenethiuram Disulfide
CAS 94-37-1
Molecular Formula C12H20N2S4
Quantity 1 g
Synonym Piperidine-1-carbothioic dithioperoxyanhydride, 1,1’-Dithiobis[1-(1-piperidinyl)-methanethione
InChI Formula InChI=1S/C12H20N2S4/c15-11(13-7-3-1-4-8-13)17-18-12(16)14-9-5-2-6-10-14/h1-10H2
SMILES S=C(SSC(=S)N1CCCCC1)N2CCCCC2
IUPAC Name piperidine-1-carbothioylsulfanyl piperidine-1-carbodithioate
Molecular Weight (g/mol) 320.56
Formula Weight 320.0509
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.