missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylchlorophosphine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7050045
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16523-54-9
C12H22ClP
232.73
MFCD00014294
AKJFBIZAEPTXIL-UHFFFAOYSA-N
chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
2734135
ClP(C1CCCCC1)C1CCCCC1
This item is not returnable. View return policy
16523-54-9
C12H22ClP
232.73
MFCD00014294
AKJFBIZAEPTXIL-UHFFFAOYSA-N
chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
2734135
ClP(C1CCCCC1)C1CCCCC1

Dicyclohexylchlorophosphine is used as pharmaceutical intermediate. It is also used in the synthesis of various phosphines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.054 g/mL
Boiling Point 97°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Linear Formula (C6H11)2PCl
Refractive Index 1.533
Quantity 1 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Reacts violently with water.
IUPAC Name chloro(dicyclohexyl)phosphane
Formula Weight 232.73
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Dicyclohexylchlorophosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'