missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexyl Phthalate, TRC

Product Code. 30382017
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382017 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382017 Supplier Toronto Research Chemicals Supplier No. TRCD4392501G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-cyclohexyl ester
CAS 84-61-7
Molecular Formula C20 H26 O4
Quantity 1 g
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dicyclohexyl ester, 1,2-Benzenedicarboxylic acid, dicyclohexyl ester (9CI), Phthalic acid, dicyclohexyl ester (6CI,8CI), DCHP, Dicyclohexyl phthalate, Edenol DCHP, Ergoplast FDC, HF 191, Howflex CP, Morflex 150, NSC 6101, Unimoll 66, Uniplex 250
InChI Formula InChI=1S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2
SMILES O=C(OC1CCCCC1)c2ccccc2C(=O)OC3CCCCC3
IUPAC Name dicyclohexyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 330.42
Formula Weight 330.1831
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.