missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclohexyl Phthalate, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-cyclohexyl ester |
| CAS | 84-61-7 |
| Molecular Formula | C20 H26 O4 |
| Quantity | 10 g |
| Synonym | 1,2-Benzenedicarboxylic acid, 1,2-dicyclohexyl ester, 1,2-Benzenedicarboxylic acid, dicyclohexyl ester (9CI), Phthalic acid, dicyclohexyl ester (6CI,8CI), DCHP, Dicyclohexyl phthalate, Edenol DCHP, Ergoplast FDC, HF 191, Howflex CP, Morflex 150, NSC 6101, Unimoll 66, Uniplex 250 |
| InChI Formula | InChI=1S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2 |
| SMILES | O=C(OC1CCCCC1)c2ccccc2C(=O)OC3CCCCC3 |
| IUPAC Name | dicyclohexyl benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 330.42 |
| Formula Weight | 330.1831 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction