missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclohexyl Phthalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-cyclohexyl ester D4 |
| CAS | 358731-25-6 |
| Molecular Formula | C20 2H4 H22 O4 |
| Quantity | 25 mg |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dicyclohexyl ester, Phthalic acid, bis-cyclohexyl ester D4, Dicyclohexyl phthalate-d4, Phthalic Acid, Bis-cyclohexyl Ester D4 (phenyl D4), 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dicyclohexyl ester, Phthalic acid, bis-cyclohexyl ester D4, Dicyclohexyl phthalate-d4 |
| InChI Formula | InChI=1S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2/i7D,8D,13D,14D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OC2CCCCC2)c(C(=O)OC3CCCCC3)c1[2H] |
| IUPAC Name | dicyclohexyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 334.44 |
| Formula Weight | 334.2082 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?