missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexyl Ketone, TRC

Product Code. 30383233
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383233 250 mg
30372200 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383233 Supplier Toronto Research Chemicals Supplier No. TRCD438870250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-60-8
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.31
Synonym Dicyclohexyl-methanone, Bicyclohexyl Ketone, Dicyclohexylmethanone, NSC 49725, NSC 60007
IUPAC Name dicyclohexylmethanone
SMILES O=C(C1CCCCC1)C2CCCCC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H22O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2
Formula Weight 194.1671
Chemical Name or Material Dicyclohexyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.