missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexano-18-crown-6, mixture of isomers, 97%

Product Code. 11488256
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488256 1 g 1g
11374509 10 g 10g
11498256 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488256 Supplier Thermo Scientific Alfa Aesar Supplier No. A15344.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16069-36-6 | C20H36O6 | 372.502 g/mol

Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

Solubility
Soluble in Toluene. Slightly soluble (13g/L, 26°C) in water.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16069-36-6
Molecular Formula C20H36O6
Molecular Weight (g/mol) 372.502
MDL Number MFCD00005099
InChI Key BBGKDYHZQOSNMU-UHFFFAOYSA-N
Synonym dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
PubChem CID 85955
SMILES C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2

Specifications

Melting Point 46°C to 53°C
Flash Point >110°C (230°F)
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 1130529
Sensitivity Hygroscopic
Merck Index 14,2602
Solubility Information Soluble in Toluene. Slightly soluble (13g/L,26°C) in water.
Formula Weight 372.5
Percent Purity 97%
Chemical Name or Material Dicyclohexano-18-crown-6, mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.