missing translation for 'onlineSavingsMsg'
Learn More

Dicumyl Peroxide, TRC

Product Code. 30137013
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137013 10 g 10g
30134082 5 g 5g
18060630 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137013 Supplier Toronto Research Chemicals Supplier No. TRCD43601310G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-43-3
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
Synonym Bis(1-methyl-1-phenylethyl) Peroxide, 14S-F1, 40B, ACEOX DCP, Active Dicumyl Peroxide, Akroform DC 40EPR, BC-FF, Bis(2-phenyl-2-propyl) Peroxide, Bis(alpha,alpha-dimethylbenzyl) Peroxide, C 3, C 3 (crosslinking agent), Cumene Peroxide;Cumyl Peroxide, D 100480, D 40C, DCP, DCP 40, DCP 40C, DCP 40KE, DCP 95R;DCP 98, DCP 99, Di-Cup, Di-Cup 40, Di-Cup 40A, Di-Cup 40C, Di-Cup 40HAF;Di-Cup 40KE, Di-Cup 40KE-U, Di-Cup 40KEU, Di-Cup 60, Di-Cup 90, Di-Cup R, Di-Cup T, Di-alpha-cumyl peroxide, Dicumene hydroperoxide, Dicumenyl peroxide;Dicumyl peroxide, Dicup 4OKE, Diisopropylbenzene peroxide, E-DCP 40P, F 40, F 40 (crosslinking agent), F 40P, Interox DCUP 1, Interox DCUP 40G, Isopropylbenzene peroxide, Kayacumyl D, Kayacumyl D 40C, Kayacumyl D 40K;Kayacumyl D 40MB, Kayacumyl D 40MB-S, Kayacumyl DCP, Kayacumyl DCP 40C;LX 500, Luperco 500-40C;
IUPAC Name 2-(2-phenylpropan-2-ylperoxy)propan-2-ylbenzene
SMILES CC(C)(OOC(C)(C)c1ccccc1)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H22O2/c1-17(2,15-11-7-5-8-12-15)19-20-18(3,4)16-13-9-6-10-14-16/h5-14H,1-4H3
Formula Weight 270.162
Chemical Name or Material Dicumyl Peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.