missing translation for 'onlineSavingsMsg'
Learn More

Dicrotophos, TRC

Product Code. 30385613
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385613 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385613 Supplier Toronto Research Chemicals Supplier No. TRCD4367201MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dicrotophos
CAS 141-66-2
Molecular Formula C8 H16 N O5 P
Quantity 1 mg
Synonym Phosphoric acid, (1E)-3-(dimethylamino)-1-methyl-3-oxo-1-propenyl dimethyl ester (9CI), Phosphoric acid, 3-(dimethylamino)-1-methyl-3-oxo-1-propenyl dimethyl ester, (E)-, Phosphoric acid, dimethyl ester, ester with 3-hydroxy-N,N-dimethylcrotonamide, (E)- (8CI), 3-(Dimethoxyphosphinyloxy)-N,N-dimethyl-cis-crotonamide, 3-Hydroxy-N,N-dimethyl-cis-crotonamide dimethyl phosphate, Bidrin, Bidrin 8, C 709, Carbicron, Carbomicron, Dicrotophos, Dimethyl 1-dimethylcarbamoyl-1-propen-2-yl phosphate, Dimethyl 2-dimethylcarbamoyl-1-methylvinyl phosphate, ENT 24,482, Ektafos, Karbicron, Oleobidrin, SD 3562, trans-Bidrin
InChI Formula InChI=1S/C8H16NO5P/c1-7(6-8(10)9(2)3)14-15(11,12-4)13-5/h6H,1-5H3/b7-6+
SMILES COP(=O)(OC)O\C(=C\C(=O)N(C)C)\C
IUPAC Name [(E)-4-(dimethylamino)-4-oxobut-2-en-2-yl] dimethyl phosphate
Molecular Weight (g/mol) 237.19
Formula Weight 237.0766
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.